siRNA Details
| Gene Symbol: ATP9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006045.2 | NP_006036.1 | 8 | 755 | ||
| XM_011528481.2 | XP_011526783.1 | 8 | 783 | |||
| GenBank mRNA | AK172802.1 | - | 6 | 444 | ||
| AK299064.1 | - | 8 | 684 | |||
| BC110591.1 | - | 4 | 247 | |||
| BC110592.1 | - | 4 | 247 | |||

ATP9A
Genomic Map