siRNA Details
| Gene Symbol: OR2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005287.1 | NP_001005287.1 | 1 | 390 | ||
| Gene Symbol: OR2A42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001802.2 | NP_001001802.2 | 1 | 390 | ||

OR2A1
Genomic Map