siRNA Details
| Gene Symbol: MCRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093767.2 | NP_001087236.1 | 3 | 217 | ||
| NM_001288798.1 | NP_001275727.1 | 4 | 353 | |||
| NM_001288799.1 | NP_001275728.1 | 3 | 217 | |||
| NM_207368.4 | NP_997251.2 | 3 | 217 | |||
| XM_011523571.2 | XP_011521873.1 | 3 | 290 | |||
| XM_011523572.2 | XP_011521874.1 | 3 | 184 | |||
| GenBank mRNA | AK000852.1 | - | 3 | 200 | ||
| AK125584.1 | - | 2 | 2269 | |||
| BC094876.1 | - | 4 | 327 | |||
| BG830426.1 | - | 3 | 218 | |||
| BU188846.1 | - | 3 | 192 | |||
| BX649156.1 | - | 3 | 215 | |||

MCRIP1
Genomic Map