siRNA Details
| Gene Symbol: MCRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093767.2 | NP_001087236.1 | 6 | 516 | ||
| NM_001288798.1 | NP_001275727.1 | 7 | 652 | |||
| NM_001288799.1 | NP_001275728.1 | 4 | 715 | |||
| NM_207368.4 | NP_997251.2 | 5 | 817 | |||
| XM_011523571.2 | XP_011521873.1 | 5 | 890 | |||
| XM_011523572.2 | XP_011521874.1 | 4 | 682 | |||
| GenBank mRNA | AK000852.1 | - | 5 | 800 | ||
| AK125584.1 | - | 5 | 2568 | |||
| BC094876.1 | - | 7 | 626 | |||
| BX649156.1 | - | 6 | 514 | |||

MCRIP1
Genomic Map