siRNA Details
| Gene Symbol: NIPAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099287.1 | NP_001092757.1 | 2 | 364 | ||
| NM_001172292.1 | NP_001165763.1 | 2 | 364 | |||
| XM_011534552.1 | XP_011532854.1 | 2 | 276 | |||
| GenBank mRNA | AK026158.1 | - | 2 | 168 | ||
| AK296972.1 | - | 2 | 364 | |||
| AK308830.1 | - | 2 | 168 | |||
| AK309242.1 | - | 2 | 276 | |||
| DB153698.1 | - | 2 | 315 | |||
| DC348015.1 | - | 2 | 364 | |||

NIPAL4
Genomic Map