siRNA Details
| Gene Symbol: LCE3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178435.3 | NP_848522.1 | 2 | 349 | ||
| XM_011509484.1 | XP_011507786.1 | 2 | 445 | |||
| GenBank mRNA | BC131716.1 | - | 2 | 349 | 40123193 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCE3E| Gene Symbol: LCE3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178435.3 | NP_848522.1 | 2 | 349 | ||
| XM_011509484.1 | XP_011507786.1 | 2 | 445 | |||
| GenBank mRNA | BC131716.1 | - | 2 | 349 | 40123193 | |