siRNA Details
| Gene Symbol: KRTAP12-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181684.2 | NP_859012.1 | 1 | 598 | ||
| GenBank mRNA | AB076362.1 | - | 1 | 577 | ||
| AJ566389.1 | - | 1 | 607 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP12-2| Gene Symbol: KRTAP12-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181684.2 | NP_859012.1 | 1 | 598 | ||
| GenBank mRNA | AB076362.1 | - | 1 | 577 | ||
| AJ566389.1 | - | 1 | 607 | |||