siRNA Details
| Gene Symbol: KRTAP10-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198694.3 | NP_941967.3 | 1 | 868 | ||
| GenBank mRNA | AB076352.1 | - | 1 | 868 | ||
| AJ566384.1 | - | 1 | 64 | |||
| BC120949.1 | - | 1 | 867 | 40115143 | ||
| BC120950.1 | - | 1 | 867 | 40115146 | ||

KRTAP10-5
Genomic Map