siRNA Details
| Gene Symbol: KRTAP12-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198697.2 | NP_941970.2 | 1 | 238 | ||
| GenBank mRNA | AB076361.1 | - | 1 | 238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP12-3| Gene Symbol: KRTAP12-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198697.2 | NP_941970.2 | 1 | 238 | ||
| GenBank mRNA | AB076361.1 | - | 1 | 238 | ||