siRNA Details
| Gene Symbol: KRTAP5-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012708.2 | NP_001012726.1 | 1 | 849 | ||
| GenBank mRNA | AJ628246.1 | - | 1 | 559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP5-3| Gene Symbol: KRTAP5-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012708.2 | NP_001012726.1 | 1 | 849 | ||
| GenBank mRNA | AJ628246.1 | - | 1 | 559 | ||