siRNA Details
| Gene Symbol: KRTAP5-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012709.1 | NP_001012727.1 | 1026 | |||
| GenBank mRNA | AB126073.1 | - | 1026 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP5-4| Gene Symbol: KRTAP5-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012709.1 | NP_001012727.1 | 1026 | |||
| GenBank mRNA | AB126073.1 | - | 1026 | |||