siRNA Details
| Gene Symbol: GOLGA2P7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027001.1 | 5 | 731 | |||
| GenBank mRNA | AK074778.1 | - | 5 | 696 | ||
| AK307326.1 | - | 6 | 788 | |||
| BC071951.1 | - | 5 | 688 | |||
| BC096759.1 | - | 5 | 688 | |||
| Gene Symbol: LOC101929479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017022812.1 | XP_016878301.1 | 5 | 718 | ||

GOLGA2P7
Genomic Map