siRNA Details
Gene Symbol: CD164L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207397.3 | NP_997280.2 | 5 | 1051 | ||
GenBank mRNA | AY358761.1 | - | 5 | 1021 | ||
BC137466.1 | - | 5 | 1010 | 9021466 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CD164L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207397.3 | NP_997280.2 | 5 | 1051 | ||
GenBank mRNA | AY358761.1 | - | 5 | 1021 | ||
BC137466.1 | - | 5 | 1010 | 9021466 |