siRNA Details
| Gene Symbol: LOC388780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287682.1 | NP_001274611.1 | 2 | 657 | ||
| XM_017027847.1 | XP_016883336.1 | 2 | 1617 | |||
| XM_017027848.1 | XP_016883337.1 | 2 | 1635 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC388780| Gene Symbol: LOC388780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287682.1 | NP_001274611.1 | 2 | 657 | ||
| XM_017027847.1 | XP_016883336.1 | 2 | 1617 | |||
| XM_017027848.1 | XP_016883337.1 | 2 | 1635 | |||