siRNA Details
| Gene Symbol: COL4A3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130105.1 | NP_001123577.1 | 7 | 1169 | ||
| NM_005713.2 | NP_005704.1 | 6 | 1070 | |||
| NM_031361.2 | NP_112729.1 | 6 | 1070 | |||
| XM_006714513.2 | XP_006714576.1 | 6 | 1070 | |||
| XM_011543090.2 | XP_011541392.1 | 6 | 1070 | |||
| XM_011543091.2 | XP_011541393.1 | 6 | 1070 | |||
| XM_017008919.1 | XP_016864408.1 | 6 | 1070 | |||
| GenBank mRNA | AF136450.1 | - | 6 | 1056 | ||
| AF232930.1 | - | 6 | 1038 | |||
| AK000604.1 | - | 2 | 90 | |||
| AK091851.1 | - | 7 | 960 | |||
| AK096854.1 | - | 7 | 1326 | |||
| AK292087.1 | - | 6 | 1070 | |||
| AY453385.1 | - | 6 | 648 | |||
| AY453386.1 | - | 6 | 648 | |||
| BC000102.2 | - | 6 | 1054 | 3505746 | ||

COL4A3BP
Genomic Map