siRNA Details
| Gene Symbol: RBAKDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015343.2 | 3 | 338 | |||
| GenBank mRNA | BC031661.1 | - | 3 | 273 | ||
| CR745855.1 | - | 3 | 231 | |||
| Gene Symbol: RBAK-RBAKDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204513.1 | NP_001191442.1 | 6 | 617 | ||

RBAKDN
Genomic Map