siRNA Details
| Gene Symbol: ARPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270439.1 | NP_001257368.1 | 3 | 599 | ||
| NM_005717.3 | NP_005708.1 | 3 | 590 | |||
| GenBank mRNA | AF006088.1 | - | 3 | 332 | ||
| AF017807.1 | - | 3 | 311 | |||
| AK095424.1 | - | 2 | 970 | |||
| AK129598.1 | - | 3 | 375 | |||
| BC057237.1 | - | 3 | 419 | 3931697 | ||
| BI758106.1 | - | 3 | 499 | |||
| CN291383.1 | - | 3 | 590 | |||

ARPC5
Genomic Map