siRNA Details
| Gene Symbol: C1QTNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207419.3 | NP_997302.2 | 3 | 471 | ||
| XM_011522483.2 | XP_011520785.1 | 3 | 554 | |||
| GenBank mRNA | AY358832.1 | - | 3 | 471 | ||
| FJ422561.1 | - | 2 | 241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1QTNF8| Gene Symbol: C1QTNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207419.3 | NP_997302.2 | 3 | 471 | ||
| XM_011522483.2 | XP_011520785.1 | 3 | 554 | |||
| GenBank mRNA | AY358832.1 | - | 3 | 471 | ||
| FJ422561.1 | - | 2 | 241 | |||