siRNA Details
| Gene Symbol: TTC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242672.1 | NP_001229601.1 | 7 | 1614 | ||
| XM_016999990.1 | XP_016855479.1 | 7 | 1686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC34| Gene Symbol: TTC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242672.1 | NP_001229601.1 | 7 | 1614 | ||
| XM_016999990.1 | XP_016855479.1 | 7 | 1686 | |||