siRNA Details
| Gene Symbol: ARPC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005720.3 | NP_005711.1 | 10 | 1306 | ||
| XM_006715825.1 | XP_006715888.1 | 10 | 1317 | |||
| XM_006715826.1 | XP_006715889.1 | 10 | 1301 | |||
| GenBank mRNA | AF006084.1 | - | 10 | 1266 | ||
| AY007111.1 | - | 8 | 1136 | |||
| BC002562.2 | - | 10 | 1239 | 3138319 | ||
| BC002988.1 | - | 10 | 1275 | 2821335 | ||
| BC007555.1 | - | 10 | 1237 | 2967684 | ||

ARPC1B
Genomic Map