siRNA Details
Gene Symbol: LOC400553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120310.1 | 1 | 246 | |||
GenBank mRNA | AK126852.1 | - | 1 | 220 | ||
CK004584.1 | - | 1 | 249 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC400553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120310.1 | 1 | 246 | |||
GenBank mRNA | AK126852.1 | - | 1 | 220 | ||
CK004584.1 | - | 1 | 249 |