siRNA Details
| Gene Symbol: LOC100996385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103794.1 | 1 | 343 | |||
| GenBank mRNA | BC042064.1 | - | 1 | 310 | ||
| HY139400.1 | - | 1 | 344 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996385| Gene Symbol: LOC100996385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103794.1 | 1 | 343 | |||
| GenBank mRNA | BC042064.1 | - | 1 | 310 | ||
| HY139400.1 | - | 1 | 344 | |||