siRNA Details
| Gene Symbol: ZNF815P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023382.1 | 4 | 579 | |||
| GenBank mRNA | AK096288.1 | - | 4 | 578 | ||
| AK291169.1 | - | 5 | 683 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF815P| Gene Symbol: ZNF815P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023382.1 | 4 | 579 | |||
| GenBank mRNA | AK096288.1 | - | 4 | 578 | ||
| AK291169.1 | - | 5 | 683 | |||