siRNA Details
| Gene Symbol: ZNF658B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003528.3 | 5 | 1280 | |||
| GenBank mRNA | AK095694.1 | - | 1 | 627 | ||
| BC137089.1 | - | 1 | 627 | 9021086 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF658B| Gene Symbol: ZNF658B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003528.3 | 5 | 1280 | |||
| GenBank mRNA | AK095694.1 | - | 1 | 627 | ||
| BC137089.1 | - | 1 | 627 | 9021086 | ||