siRNA Details
| Gene Symbol: RNF223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205252.1 | NP_001192181.1 | 2 | 484 | ||
| XM_017001286.1 | XP_016856775.1 | 2 | 484 | |||
| GenBank mRNA | AK296576.1 | - | 2 | 484 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF223| Gene Symbol: RNF223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205252.1 | NP_001192181.1 | 2 | 484 | ||
| XM_017001286.1 | XP_016856775.1 | 2 | 484 | |||
| GenBank mRNA | AK296576.1 | - | 2 | 484 | ||