siRNA Details
| Gene Symbol: TSPAN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005723.3 | NP_005714.2 | 1, 2 | 523 | ||
| XM_005262680.1 | XP_005262737.1 | 1, 2 | 523 | |||
| GenBank mRNA | AF065389.1 | - | 1, 2 | 435 | ||
| AK315723.1 | - | 1, 2 | 490 | |||
| BC009704.1 | - | 1, 2 | 421 | 3895933 | ||
| CR541809.1 | - | 1, 2 | 88 | |||
| CR541829.1 | - | 1, 2 | 88 | |||

TSPAN5
Genomic Map