siRNA Details
| Gene Symbol: KRTAP5-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012503.1 | NP_001012521.1 | 1 | 646 | ||
| GenBank mRNA | AB126076.1 | - | 1 | 646 | ||
| AJ628243.1 | - | 1 | 232 | |||
| BC136967.1 | - | 1 | 635 | 9020959 | ||
| BC136968.1 | - | 1 | 635 | 9020960 | ||

KRTAP5-7
Genomic Map