siRNA Details
| Gene Symbol: SMIM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253790.1 | NP_001240719.1 | 5 | 746 | ||
| NM_001253791.1 | NP_001240720.1 | 6 | 553 | |||
| NM_001253793.1 | NP_001240722.1 | 4 | 318 | |||
| NM_001253794.1 | NP_001240723.1 | 4 | 333 | |||
| XM_011522499.2 | XP_011520801.1 | 5 | 763 | |||
| XM_011522500.2 | XP_011520802.1 | 6 | 881 | |||
| XM_011522501.2 | XP_011520803.1 | 5 | 571 | |||
| GenBank mRNA | BC022385.2 | - | 3 | 262 | ||
| BC035868.1 | - | 5 | 746 | |||
| BC048326.1 | - | 6 | 553 | |||
| BC127921.1 | - | 1 | 503 | |||
| BC148245.1 | - | 2 | 414 | |||
| BI767985.1 | - | 4 | 318 | |||
| CK903133.1 | - | 3 | 243 | |||

SMIM22
Genomic Map