siRNA Details
| Gene Symbol: LINC00851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034167.1 | 2 | 691 | |||
| GenBank mRNA | BC033985.1 | - | 2 | 696 | 5295734 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00851| Gene Symbol: LINC00851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034167.1 | 2 | 691 | |||
| GenBank mRNA | BC033985.1 | - | 2 | 696 | 5295734 | |