siRNA Details
| Gene Symbol: FAR2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026758.2 | 11 | 2865 | |||
| GenBank mRNA | AK095330.1 | - | 1 | 3486 | ||
| AK125921.1 | - | 11 | 2866 | |||
| BC132694.1 | - | 6 | 562 | |||
| BC132696.1 | - | 6 | 562 | |||
| BC144438.1 | - | 6 | 563 | 9052961 | ||
| EU668357.1 | - | 2 | 1085 | |||

FAR2P1
Genomic Map