siRNA Details
| Gene Symbol: TSFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172696.1 | NP_001166167.1 | 6, 7 | 699 | ||
| NM_005726.5 | NP_005717.3 | 5, 6 | 636 | |||
| GenBank mRNA | AF110399.1 | - | 5, 6 | 582 | ||
| AK295323.1 | - | 7, 8 | 868 | |||
| AK313453.1 | - | 5, 6 | 620 | |||
| BC022862.1 | - | 6, 7 | 672 | 4076880 | ||
| L37936.1 | - | 4, 5 | 487 | |||

TSFM
Genomic Map