siRNA Details
| Gene Symbol: FAM90A7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046343.1 | 3 | 1488 | |||
| GenBank mRNA | AK301810.1 | - | 3 | 1489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM90A7P| Gene Symbol: FAM90A7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046343.1 | 3 | 1488 | |||
| GenBank mRNA | AK301810.1 | - | 3 | 1489 | ||