siRNA Details
| Gene Symbol: FAM90A7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046343.1 | 3 | 1764 | |||
| Gene Symbol: FAM90A25P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073395.1 | 4 | 1819 | |||
| GenBank mRNA | AL832996.1 | - | 4 | 1818 | ||

FAM90A7P
Genomic Map