siRNA Details
| Gene Symbol: RARB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290216.2 | NP_001277145.1 | 4 | 533 | ||
| GenBank mRNA | AK299756.1 | - | 1 | 108 | ||
| DC376623.1 | - | 1 | 108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RARB| Gene Symbol: RARB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290216.2 | NP_001277145.1 | 4 | 533 | ||
| GenBank mRNA | AK299756.1 | - | 1 | 108 | ||
| DC376623.1 | - | 1 | 108 | |||