siRNA Details
| Gene Symbol: TRIM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006778.3 | NP_006769.2 | 3 | 662 | ||
| NM_052828.2 | NP_439893.2 | 3 | 662 | |||
| XM_011514221.1 | XP_011512523.1 | 5 | 910 | |||
| XM_011514222.2 | XP_011512524.1 | 5 | 1144 | |||
| XM_011514223.2 | XP_011512525.1 | 6 | 1025 | |||
| XM_011514225.1 | XP_011512527.1 | 5 | 910 | |||
| GenBank mRNA | AF220122.1 | - | 3 | 660 | ||
| AF220123.1 | - | 3 | 660 | |||
| BC093926.1 | - | 3 | 640 | 7939771 | ||
| BC113419.1 | - | 3 | 640 | 8322471 | ||

TRIM10
Genomic Map