siRNA Details
| Gene Symbol: BOLA2-SMG1P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320622.1 | NP_001307551.1 | 1 | 581 | ||
| NM_001320623.1 | NP_001307552.1 | 1 | 581 | |||
| NM_001320624.1 | NP_001307553.1 | 1 | 581 | |||
| NM_001320625.1 | NP_001307554.1 | 1 | 581 | |||
| NM_001320627.1 | NP_001307556.1 | 1 | 581 | |||
| NR_135316.1 | 1 | 581 | ||||
| GenBank mRNA | AF271775.1 | - | 1 | 177 | ||
| BC130401.1 | - | 1 | 268 | 40146432 | ||
| BC130403.1 | - | 1 | 268 | 40146434 | ||
| Gene Symbol: BOLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031827.2 | NP_001026997.1 | 1 | 581 | ||
| NM_001320579.1 | NP_001307508.1 | 1 | 581 | |||
| GenBank mRNA | BC022832.1 | - | 1 | 581 | 4647797 | |
| BC144688.1 | - | 1 | 268 | 9053222 | ||
| Gene Symbol: BOLA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039182.2 | NP_001034271.1 | 1 | 581 | ||
| NM_001320607.1 | NP_001307536.1 | 1 | 581 | |||
| GenBank mRNA | BC137532.1 | - | 1 | 268 | 9021532 | |
| BC137534.1 | - | 1 | 268 | 9021534 | ||

BOLA2-SMG1P6
Genomic Map