siRNA Details
| Gene Symbol: TRIM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006778.3 | NP_006769.2 | 1, 2 | 513 | ||
| NM_052828.2 | NP_439893.2 | 1, 2 | 513 | |||
| XM_011514221.1 | XP_011512523.1 | 3, 4 | 761 | |||
| XM_011514222.2 | XP_011512524.1 | 3, 4 | 995 | |||
| XM_011514223.2 | XP_011512525.1 | 4, 5 | 876 | |||
| XM_011514225.1 | XP_011512527.1 | 3, 4 | 761 | |||
| GenBank mRNA | AF220122.1 | - | 1, 2 | 511 | ||
| AF220123.1 | - | 1, 2 | 511 | |||
| BC093926.1 | - | 1, 2 | 491 | 7939771 | ||
| BC113419.1 | - | 1, 2 | 491 | 8322471 | ||

TRIM10
Genomic Map