siRNA Details
| Gene Symbol: FBRSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005266181.3 | XP_005266238.1 | 3 | 2312 | ||
| GenBank mRNA | BC013284.2 | - | 1 | 1244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBRSL1| Gene Symbol: FBRSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005266181.3 | XP_005266238.1 | 3 | 2312 | ||
| GenBank mRNA | BC013284.2 | - | 1 | 1244 | ||