siRNA Details
| Gene Symbol: C2orf27B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214461.2 | NP_999626.1 | 3 | 917 | ||
| GenBank mRNA | BC043584.1 | - | 3 | 917 | 5229449 | |
| Gene Symbol: C2orf27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013310.3 | NP_037442.3 | 4 | 811 | ||
| GenBank mRNA | AF038169.1 | - | 4 | 811 | ||
| BC112189.1 | - | 2 | 431 | 8327657 | ||
| Gene Symbol: LOC105372731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028519.1 | XP_016884008.1 | 2 | 628 | ||

C2orf27B
Genomic Map