siRNA Details
| Gene Symbol: MAP1LC3B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085481.1 | NP_001078950.1 | 2 | 629 | ||
| XM_011538652.2 | XP_011536954.1 | 2 | 802 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP1LC3B2| Gene Symbol: MAP1LC3B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085481.1 | NP_001078950.1 | 2 | 629 | ||
| XM_011538652.2 | XP_011536954.1 | 2 | 802 | |||