siRNA Details
| Gene Symbol: FAM163B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080515.2 | NP_001073984.1 | 1 | 91 | ||
| XM_005272203.4 | XP_005272260.1 | 2 | 968 | |||
| XM_005272204.4 | XP_005272261.1 | 4 | 416 | |||
| XM_006717233.3 | XP_006717296.1 | 2 | 378 | |||
| XM_011518912.2 | XP_011517214.1 | 2 | 912 | |||
| XM_011518913.2 | XP_011517215.1 | 2 | 2001 | |||
| XM_017015017.1 | XP_016870506.1 | 2 | 406 | |||
| XM_017015018.1 | XP_016870507.1 | 2 | 674 | |||
| GenBank mRNA | BC146946.1 | - | 1 | 91 | 9056874 | |
| BC146961.1 | - | 1 | 91 | 9056889 | ||

FAM163B
Genomic Map