siRNA Details
| Gene Symbol: RNF222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146684.2 | NP_001140156.1 | 3 | 433 | ||
| XM_011523978.2 | XP_011522280.1 | 3 | 410 | |||
| XM_011523979.2 | XP_011522281.1 | 3 | 518 | |||
| XM_011523980.2 | XP_011522282.1 | 2 | 302 | |||
| XM_011523981.2 | XP_011522283.1 | 2 | 246 | |||

RNF222
Genomic Map