siRNA Details
| Gene Symbol: RASGRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128602.1 | NP_001122074.1 | 16 | 2372 | ||
| NM_001306086.1 | NP_001293015.1 | 15 | 1986 | |||
| NM_005739.3 | NP_005730.2 | 17 | 2477 | |||
| XM_005254114.2 | XP_005254171.1 | 16 | 2313 | |||
| XM_011521151.2 | XP_011519453.1 | 16 | 2080 | |||
| XM_017021860.1 | XP_016877349.1 | 17 | 2404 | |||
| GenBank mRNA | AB208848.1 | - | 10 | 1498 | ||
| AF081195.1 | - | 17 | 2402 | |||
| AF081197.1 | - | 16 | 2256 | |||
| AF106071.1 | - | 17 | 2311 | |||
| AK295097.1 | - | 16 | 2341 | |||
| AK314091.1 | - | 17 | 2477 | |||
| AK316067.1 | - | 17 | 2405 | |||
| AY634315.2 | - | 14 | 1662 | |||
| AY858556.2 | - | 15 | 1815 | |||
| AY954625.4 | - | 16 | 2201 | |||
| AY966005.1 | - | 15 | 2056 | |||
| BC109296.1 | - | 17 | 2349 | 40034427 | ||
| BC109297.1 | - | 17 | 2349 | 40034428 | ||
| BU431038.1 | - | 2 | 140 | |||

RASGRP1
Genomic Map