siRNA Details
| Gene Symbol: RPS14P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077246.1 | 1 | 689 | |||
| GenBank mRNA | BC072682.1 | - | 1 | 689 | ||
| BU176905.1 | - | 1 | 657 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS14P3| Gene Symbol: RPS14P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_077246.1 | 1 | 689 | |||
| GenBank mRNA | BC072682.1 | - | 1 | 689 | ||
| BU176905.1 | - | 1 | 657 | |||