siRNA Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005740.2 | NP_005731.1 | 4 | 493 | ||
| GenBank mRNA | AK223349.1 | - | 4 | 480 | ||
| AK290566.1 | - | 4 | 446 | |||
| AL035366.1 | - | 4 | 486 | |||
| AY453399.1 | - | 284 | ||||
| BC002968.1 | - | 4 | 460 | 3543962 | ||
| BT009847.1 | - | 3 | 265 | |||
| BX473356.1 | - | 4 | 498 | |||
| CR456487.1 | - | 3 | 350 | |||
| CR542177.1 | - | 3 | 265 | |||
| CR542195.1 | - | 3 | 265 | |||

DNAL4
Genomic Map