siRNA Details
| Gene Symbol: ZNF263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005741.4 | NP_005732.2 | 3 | 948 | ||
| XM_006720832.1 | XP_006720895.1 | 3 | 1055 | |||
| XM_006720834.3 | XP_006720897.1 | 2 | 1071 | |||
| XM_011522344.1 | XP_011520646.1 | 3 | 1055 | |||
| XM_017022850.1 | XP_016878339.1 | 2 | 494 | |||
| GenBank mRNA | AK312421.1 | - | 3 | 930 | ||
| BC008805.2 | - | 3 | 946 | 3945548 | ||
| D88827.1 | - | 3 | 908 | |||

ZNF263
Genomic Map