siRNA Details
| Gene Symbol: LOC101929060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027550.1 | XP_016883039.1 | 1209 | |||
| Gene Symbol: MIER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB033019.1 | - | 5358 | |||

LOC101929060
Genomic Map