siRNA Details
| Gene Symbol: GOLGA8S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038843.1 | 18 | 2878 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GOLGA8S| Gene Symbol: GOLGA8S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038843.1 | 18 | 2878 | |||