siRNA Details
| Gene Symbol: LOC101928697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126054.1 | 3 | 252 | |||
| GenBank mRNA | HY007484.1 | - | 3 | 253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928697| Gene Symbol: LOC101928697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126054.1 | 3 | 252 | |||
| GenBank mRNA | HY007484.1 | - | 3 | 253 | ||