siRNA Details
| Gene Symbol: RMND5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005265971.4 | XP_005266028.1 | 1 | 102 | ||
| XM_017009727.1 | XP_016865216.1 | 1 | 102 | |||
| XM_017009728.1 | XP_016865217.1 | 1 | 102 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RMND5B| Gene Symbol: RMND5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005265971.4 | XP_005266028.1 | 1 | 102 | ||
| XM_017009727.1 | XP_016865216.1 | 1 | 102 | |||
| XM_017009728.1 | XP_016865217.1 | 1 | 102 | |||